Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551786 Supplier Invitrogen™ Supplier No. PA551786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEQGRKASTA FEQTMFLSRA LEEIQTCNDI HNFLKQTGTF SNDSCEKLLL LYEFEVRAKL NDPLLESFLE SVWELPHLET KTFETIAIIA MEKPAHYPLI ALKALKKALL LYKKEEPIDI SQYSKCMHNL V Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 60%.

This gene is X-linked and is expressed in only male germ cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX11
Gene Accession No. Q8IYF3
Gene Alias 4930565P14Rik; BB024231; Protein ZIP4 homolog; RGD1560239; SPGFX2; testis expressed 11; testis expressed gene 11; testis expressed sequence 11; testis-expressed protein 11; testis-expressed sequence 11 protein; testis-expressed sequence 11 protein; testis-expressed protein 11; Tex11; TGC1; TSGA3; ZIP4H
Gene Symbols TEX11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEX11. Recombinant protein control fragment (Product #RP-92193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.