Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558326 Supplier Invitrogen™ Supplier No. PA558326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKEINLMLST YAKLLSERAA VDASYIDEID ELFKEANAIE NFLIQKREFL RQRFTVIANT LHR Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

This gene is similar to a mouse gene that is expressed in the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX12
Gene Accession No. Q9BXU0
Gene Alias 5730518K06Rik; testis expressed 12; testis expressed gene 12; testis expressed sequence 12; testis exprtessed gene 12; Testis-expressed protein 12; testis-expressed sequence 12 protein; testis-expressed sequence 12 protein variant 1; testis-expressed sequence 12 protein variant 2; testis-expressed sequence 12 protein variant 3; Tex12
Gene Symbols TEX12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEX12. Recombinant protein control fragment (Product #RP-97408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56158
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.