Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX13A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584891 Supplier Invitrogen™ Supplier No. PA584891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TISPPQATVT APVPPQLPSD WEAFDTSLWS DGGPHRIDHQ EHPRDRRYSE PHQQRPPVYR RPGDWDCPWC NAVNFSRRDT CFDC Highest antigen sequence indentity to the following orthologs: Rat - 26%, Mouse - 43%.

This gene is similar to a mouse gene that is expressed in the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX13A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX13A
Gene Accession No. Q9BXU3
Gene Alias 1700025D03Rik; testis expressed 13A; Testis-expressed protein 13A; testis-expressed sequence 13A protein; TEX13A
Gene Symbols TEX13A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEX13A. Recombinant protein control fragment (Product #RP-108256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.