Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEX14 Monoclonal Antibody (1A11D6), CoraLite594, Proteintech
SDP

Catalog No. 502334751 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4751 Supplier Proteintech Group Inc Supplier No. CL59467982100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TEX14 Monoclonal antibody specifically detects TEX14 in Human, Mouse, Rabbit, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TEX14
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A11D6
Concentration 1 mg/mL
Conjugate CoraLite 594
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. Q8IWB6
Gene Alias C85585; CT113; SGK307; TEX14
Host Species Mouse
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100339930
Target Species Rabbit
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.