Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544140 Supplier Invitrogen™ Supplier No. PA544140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TPDGEYFYSS TAQENLALET SSPIEEDFEG IQGAFAQPQV SGEEKFQMRK Sequence homology: Cow: 86%; Dog: 79%; Horse: 82%; Human: 100%; Pig: 85%; Rabbit: 79%; Rat: 79%.

This gene is similar to a mouse gene that is expressed in the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEX14
Gene Accession No. Q8IWB6
Gene Alias C85585; cancer/testis antigen 113; CT113; Inactive serine/threonine-protein kinase TEX14; LOW QUALITY PROTEIN: inactive serine/threonine-protein kinase TEX14; protein kinase-like protein SgK307; SGK307; Sugen kinase 307; testis expressed 14; testis expressed 14, intercellular bridge forming factor; testis expressed gene 14; testis expressed sequence 14; testis-expressed protein 14; Testis-expressed sequence 14; Testis-expressed sequence 14 protein; Tex14; Unknown (protein for MGC:133568)
Gene Symbols TEX14
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TEX14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56155
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.