Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557824 Supplier Invitrogen™ Supplier No. PA557824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVYKRSMTEG STVNTEYKNQ KNQISEESCL NEKIITTNLI DSHLSTKNTT TESVPLKNTV SNPLNKREKK GEIKVSKDSQ SDLTLHSEIA YISKPGILGV NHTPILPAHS ETCKVPTLL Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 38%.

Required during spermatogenesis for normal chromosome synapsis and meiotic recombination in germ cells. Necessary for formation of DMC1 and RAD51 foci on meiotic chromosomes, suggesting a specific role in DNA double-stranded break repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX15
Gene Accession No. Q9BXT5
Gene Alias 2210014E14Rik; AL022622; AU022940; cancer/testis antigen 42; CT42; testis expressed 15; testis expressed gene 15; Testis-expressed protein 15; testis-expressed sequence 15 protein; TEX15
Gene Symbols TEX15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEX15. Recombinant protein control fragment (Product #RP-97580).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.