Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tex19.1 Monoclonal Antibody (678826)
GREENER_CHOICE

Catalog No. PIMA524308
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524308 Supplier Invitrogen™ Supplier No. MA524308
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tex19.1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 678826
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Tex19.1
Gene Accession No. Q99MV2
Gene Alias 2410081M02Rik; mTex19.1; testis expressed 19; testis expressed gene 19; testis expressed gene 19.1; Testis-expressed protein 19; Testis-expressed protein 19.1; testis-expressed protein 19A; testis-expressed sequence 19 protein; testis-expressed sequence 19A; Tex19; Tex19.1; Tex19a
Gene Symbols Tex19.1
Host Species Rat
Immunogen E. coli-derived recombinant mouse TEX19.1 Arg189-Pro351.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73679
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.