Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tex19.1, Mouse anti-Mouse, Clone: 7TEX-1F11, Millipore Sigma™

Catalog No. MABC1596100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1596100 100 μg
MABC159625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1596100 Supplier MilliporeSigma Supplier No. MABC1596100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tex19.1, clone 7TEX-1F11, Cat. No. MABC1596, is a mouse monoclonal antibody that detects Testis-expressed protein 19.1 and has been tested for use in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Testis-expressed protein 19.1 (UniProt: Q99MV2; also known as mTex19.1, Testis-expressed protein 19A) is encoded by the Tex19.1 (also known as Tex19, Tex19a) gene (Gene ID: 73679) in murine species. Tex19.1 resides in the germline to repress transposable genetic elements and maintain genomic stability. It is expressed in testis, placenta, ovary, and in pluripotent stem cells. In testis its expression is highest in mitotic spermatogonia and decreases as spermatocytes progress through meiosis. Tex19.1 expression is reported to be down-regulated by Deleted in azoospermia-like (DAZL) protein, which binds to 3'UTR of Tex19.1 mRNAs and repress its translation. Tex19.1 is required during spermatogenesis and placenta development where it participates in the repression of retrotransposable elements and prevents their mobilization. Tex19.1, along with its paralog, Tex19.2, is shown to collaborate with the Piwi-interacting RNA (piRNA) pathway, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins. Embryos with mutations in Tex19.1 gene exhibit intra-uterine growth retardation and have small placentas due to a reduction in the number of spongiotrophoblast, glycogen trophoblast and sinusoidal trophoblast giant cells. Loss of Tex19.1 is shown to cause growth defect and early post-natal lethality. Male animals with deficiency of both Tex19.1 and Tex19.2 display impaired spermatogenesis, small testes, and are infertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tex19.1
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7TEX-1F11
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Tex19.1 in embryonic mouse brain and embryonic mouse gut tissue sections.Immunocytochemistry Analysis: A representative lot detected Tex19.1 in Immunocytochemistry applications (Tarabay, Y., et. al. (2013). Hum Reprod. 28(8):2201-14).ELISA Analysis: A representative lot detected Tex19.1 in ELISA applications (Tarabay, Y., et. al. (2013). Hum Reprod. 28(8):2201-14).Immunoprecipitation Analysis: A representative lot immunoprecipitated Tex19.1 in Immunoprecipitation applications (Tarabay, Y., et. al. (2017). J Cell Sci. 130(8):1463-1474).Western Blotting Analysis: A representative lot detected Tex19.1 in Western Blotting applications (Tarabay, Y., et. al. (2013). Hum Reprod. 28(8):2201-14;Tarabay, Y., et. al. (2017). J Cell Sci. 130(8):1463-1474).
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Testis-expressed protein 19.1;mTex19.1;Testis-expressed protein 19A
Gene Symbols Tex19.1;Tex19;Tex19a
Host Species Mouse
Immunogen His-tagged full-length recombinant mouse Testis-expressed protein 19.1 (Tex 19).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_082878
Test Specificity Clone 7TEX-1F11 detects murine Testis-expressed protein 19.1 (Tex 19).
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.