Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548099
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548099 Supplier Invitrogen™ Supplier No. PA548099
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 2% cross-reactivity with recombinant human TEX11 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TEX19
Gene Accession No. Q8NA77
Gene Alias FLJ35767; testis expressed 19; Testis-expressed protein 19; testis-expressed sequence 19 protein; TEX19
Gene Symbols TEX19
Host Species Sheep
Immunogen E. coli-derived recombinant human TEX19 Met1-Ser164.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400629
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.