Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEX2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$158.00 - $487.50

Specifications

Antigen TEX2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15997720
SDP
View Documents
Novus Biologicals
NBP15997720UL
20 μL
Each for $158.00
Only null left
Add to Cart
 
NBP159977
SDP
View Documents
Novus Biologicals
NBP159977
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

TEX2 Polyclonal specifically detects TEX2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TEX2
Polyclonal
Rabbit
Q8IWB9
55852
Synthetic peptides corresponding to TEX2(testis expressed 2) The peptide sequence was selected from the N terminal of TEX2. Peptide sequence KSLSTEVEPKESPHPARHRHLMKTLVKSLSTDTSRQESDTVSYKPPDSKL.
Primary
Western Blot
Unconjugated
RUO
HT008, KIAA1738DKFZp781G0721, testis expressed 2, testis expressed sequence 2, testis-expressed sequence 2 protein, TMEM96, transmembrane protein 96
TEX2
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.