Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554255
Description
Immunogen sequence: QLLSKIIGEE SGSHRPKALS SSASELSNLS SLNGHLESNN NYSIKEEECD SEGDGYGSDS NIPRSDHPKS TGEPTREIEL KSSQGSSLKD L Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
TEX2 is a multi-pass membrane protein. The exact function of TEX2 remains unknown.
Specifications
TEX2 | |
Polyclonal | |
Unconjugated | |
TEX2 | |
4930568E07Rik; AI553404; Def-5; HT008; Kiaa1738; Taz4; testis expressed 2; testis expressed gene 2; testis expressed sequence 2; testis-expressed protein 2; testis-expressed sequence 2 protein; testis-expressed sequence 2 protein; testis-expressed protein 2; Tex2; TMEM96; transmembrane protein 96 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55852 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IWB9 | |
TEX2 | |
Recombinant protein corresponding to Human TEX2. Recombinant protein control fragment (Product #RP-90964). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction