Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEX261 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981944
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1944 Supplier Bioss Supplier No. BS12105R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen TEX261
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TEX261
Gene Accession No. Q6UWH6
Gene Alias 3110001O07Rik; AA409339; AI480706; AL033351; Protein TEX261; TEG-261; testis expressed 261; testis expressed gene 261; testis expressed sequence 261; Testis-expressed protein 261; testis-expressed sequence 261 protein; Tex261; UNQ1882/PRO4325
Gene Symbols TEX261
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TEX261.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113419
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.