Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEX264 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107036
Description
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.
The specific function of TEX264 is not yet known.
Specifications
TEX264 | |
Polyclonal | |
Unconjugated | |
Tex264 | |
Putative secreted protein Zsig11; TEG-264; testis expressed 264; testis expressed gene 264; Testis-expressed protein 264; testis-expressed sequence 264 protein; Tex264; UNQ337/PRO536; ZSIG11 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q9Y6I9 | |
Tex264 | |
Synthetic peptide directed towards the middle region of human TEX264 (aa 143-193). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction