Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEX35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113002
Description
TEX35 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The function of this protein remains unknown.
Specifications
TEX35 | |
Polyclonal | |
Unconjugated | |
TEX35 | |
C1orf49; testis expressed 35; testis-expressed protein 35; testis-expressed sequence 35 protein; Testis-Specific Conserved gene 24kDa; TEX35; TSC24; uncharacterized protein C1orf49 homolog | |
Rabbit | |
Protein G | |
RUO | |
84066 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q5T0J7 | |
TEX35 | |
Recombinant Human Testis-expressed protein 35 protein (1-215AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction