Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEX35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545990
Description
Peptide sequence: CLLCALKNNY NRGNIPSEAS GLYKGGEEPV TTQPSVGHAV PAPKSQTEGR Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
TEX35 | |
Polyclonal | |
Unconjugated | |
TEX35 | |
C1orf49; testis expressed 35; testis-expressed protein 35; testis-expressed sequence 35 protein; Testis-Specific Conserved gene 24kDa; TEX35; TSC24; uncharacterized protein C1orf49 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84066 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5T0J7 | |
TEX35 | |
Synthetic peptide directed towards the C-terminal of human C1orf49 (aa 184-233). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction