Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TEX36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571033
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Rabbit: 100%; Rat: 93%.
TEX36 is a protein coding gene.
Specifications
TEX36 | |
Polyclonal | |
Unconjugated | |
TEX36 | |
bA383C5.1; C10orf122; C26H10orf122; C28H10orf122; hypothetical gene supported by BC079265; testis expressed 36; testis-expressed protein 36; testis-expressed sequence 36 protein; testis-expressed sequence 36 protein; testis-expressed protein 36; Tex36; Unknown (protein for MGC:133583) | |
Rabbit | |
Affinity chromatography | |
RUO | |
387718 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VZQ5 | |
TEX36 | |
synthetic peptide directed towards the middle region of human TEX36. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction