Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139897 Supplier Invitrogen™ Supplier No. PA5139897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GFFPSQEHEA TREDERKFTS TCHFTYPASH DLHLAQGDPN QVLQSADFPC LVDPKHQPAA EMAKGYLLLP GCPCLHCH Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 58%.

C2orf51, also named as TEX37 and TSC21, is a protein with calculated MW 21 kDa. We always detect 35 kDa in our WB detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX37
Applications Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEX37
Gene Accession No. Q96LM6
Gene Alias 1700011F03Rik; C2orf51; protein TSC21; testis expressed 37; testis tissue sperm-binding protein Li 93mP; testis-expressed sequence 37 protein; Testis-Specific Conserved gene 21kDa; testis-specific conserved protein of 21 kDa; TEX37; Tsc21
Gene Symbols TEX37
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TEX37. Recombinant protein control fragment (Product #RP-109200).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.