Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEX55 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23799125UL
Description
TEX55 Polyclonal specifically detects TEX55 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
C3orf30 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q96M34 | |
C3ORF30 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QAERRTSGQIDGRLAMPSDQRGSRQTDHRMAGQSERRASEQMDRRMSGEAERRTSEQITHRLSKLSERRPSVQIDSGSSVPSDQSP | |
25 μL | |
Protein Kinase | |
152405 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
uncharacterized protein C3orf30, chromosome 3 open reading frame 30 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction