Description
Aspirin tablets. Easy-to-swallow coated tablets relieve pain, inflammation, and fever. Each tablet contains 325mg aspirin; no sugar, salt, lactose, or caffeine. Two tablets per packet, 100 or 250 packets per dispenser pack.Extra-strength pain relief tablets. For relief of symptoms associated with headaches, menstrual cramps, muscular aches and pains, and fever. Each tablet contains 162mg aspirin, 110mg acetaminophen, 152mg salicyamide, and 32.4mg caffeine. Two tablets per packet, 50 or 250 packets per dispenser pack.Tri-buffered aspirin tablets. Similar to regular aspirin yet gentle on the stomach. Each tablet contains 325mg aspirin buffered with magnesium oxide. Two tablets per packet, 50 or 250 packets per dispenser pack.
Specifications
Specifications
| Active Ingredients | Acetaminophen (110 mg), Aspirin (162 mg), Salicylamide (152 mg), Caffeine (32.4 mg) |
| Format | Film-coated Tablet |
| Quantity | 100/Pk. |