Learn More
Valutek™ Textured Nylon Wiper, Cold Knife Cut
Catalog No. p-7162574
Click to view available options
Dimensions (L x W):
9 x 9 in.
12 x 12 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
Constructed from 100% continuous filament nylon in a double-knit, no-run, interlock pattern. This clean and absorbent wiper offers excellent softness and superior strength.
  • 100% continuous filament, double-knit nylon fiber
  • Cold knife cut edge
  • Excellent tensile strength, elasticity and durability
  • Low levels of particulate and extractable counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Laundered and packaged in a cleanroom, all Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 100-1000 (ISO 5-6) critical environment
  • The tough and abrasive surface of nylon makes it a great selection for cleaning stubborn contaminants
Specifications
Material Nylon
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.