Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
SDP
Catalog No. p-3557201
Click to view available options
Product Type:
Cleanroom Mop
Mop
Replacement Mop Covers
Includes
Two handles (18 and 24 in. long), one polyester foam pad, and six TechniCloth™ polyester mop covers. Mop with telescoping handle includes six polyester mop covers.
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
  • Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
  • Made from autoclavable Celcon™ plastic
  • Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Product Type Replacement Mop Covers
Quantity 150/Cs.
For Use With (Application) For Cleanroom Class 4 to 8
Material Polyester
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.