Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
SDP
Catalog No. 19166872
Click to view available options
Color:
Blue
Gray
White
Dimensions (L x W):
18 x 36 in.
36 x 18 in.
45 x 25 in.
46 x 36 in.
Apply directly to floor
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Color Gray
Dimensions (L x W) 45 x 25 in.
Quantity 60 Sheets/Mat
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.