Learn More
Texwipe™ TOC Cleaning Validation Kits
SDP
Catalog No. 18315210 Shop All Texwipe Products
Change view
Click to view available options
Product Type:
Validation Kit
Validation Kit, Long Handle Swab
For Use With (Application):
Sampling 12 Production Areas
Sampling 72 Production Areas
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
18-315-210 Validation Kit, Long Handle Swab Sampling 12 Production Areas
19-130-3721 Validation Kit Sampling 72 Production Areas
19-130-3720 Validation Kit Sampling 12 Production Areas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-315-210 Supplier Texwipe™ Supplier No. TX3343
This item is not returnable. View return policy
Includes
Vials, swabs, and labels for sampling 12 or 72 production areas, and container for sample transport.
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
  • Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
  • Prewashed vials have one-piece cap and septa design for easy sampling and analysis
  • Polyethylene overcap protects septum surface from contamination until time of analysis
  • Sloped shoulder design minimizes headspace for reduced background TOC level
  • Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
Sampling Swab
  • Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
  • Double layered, double knit polyester head entraps contaminants in knit structure during sampling
  • Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
  • Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
  • 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Certifications
Certificate of Analysis included on each box.
Specifications
Product Type Validation Kit, Long Handle Swab
For Use With (Application) Sampling 12 Production Areas
Includes 12 40 mL clear vials bonded septa cap, 24 Low TOC Alpha™ Sampling Swab with Long Handle, 12 blank vial labels
Quantity 18/Cs.