Learn More
TF Mouse anti-Human, Clone: OTI5G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500848

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.Specifications
TF | |
Monoclonal | |
Unconjugated | |
TF | |
HEL-S-71p, PRO1557, PRO2086, TFQTL1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7018 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI5G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02787 | |
TF | |
Full length human recombit protein of human TF produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.