Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP2A Mouse anti-Human, Monkey, Mouse, Rat, Clone:7D2B5, Abnova™

Catalog No. 89333154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-154 Supplier Abnova Corporation Supplier No. MAB16704
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human TFAP2A.

AP2-alpha is a 52-kD retinoic acid-inducible and developmentally regulated activator of transcription that binds to a consensus DNA-binding sequence CCCCAGGC in the SV40 and metallothionein (MIM 156350) promoters.[supplied by OMIM]

Specifications

Antigen TFAP2A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7D2B5
Conjugate Unconjugated
Description transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene TFAP2A
Gene Alias AP-2, AP-2alpha, AP2TF, BOFS, TFAP2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-100 of human TFAP2A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 7020
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.