Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFAP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110476
Description
Immunogen sequence: MHSPPRDQAA IMLWKLVENV KYEDIYEDRH DGVPSHSSRL SQLGSVSQGP YSSAPPLSHT PSSDFQPPYF PPPYQPLPYH QSQDPYSHVN DPYSLNPLHQ.
This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives.
Specifications
| TFAP2B | |
| Polyclonal | |
| Unconjugated | |
| Tfap2b | |
| activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 beta; activating enhancer-binding protein 2-beta; AI606113; AP-2(beta); AP-2B; AP2-B; AP2-beta; E130018K07Rik; Tcfap2b; Tfap2b; transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); transcription factor AP-2-beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7021 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92481 | |
| Tfap2b | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TFAP2B (Q92481). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction