Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP2C Mouse anti-Human, Clone:3A11A5, Abnova™

Catalog No. 89334355 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-355 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-355 Supplier Abnova Corporation Supplier No. MAB17907
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human TFAP2C.

The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube. [provided by RefSeq]

Specifications

Antigen TFAP2C
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A11A5
Conjugate Unconjugated
Description transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma)
Dilution ELISA (1:10000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene TFAP2C
Gene Alias AP2-GAMMA, ERF1, TFAP2G, hAP-2g
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 341-450 of human TFAP2C.
Quantity 100 μg
Gene ID (Entrez) 7022
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.