Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFAP2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100768 Supplier Invitrogen™ Supplier No. PA5100768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TFAP2D.

Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFAP2D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TFAP2D
Gene Accession No. Q7Z6R9, Q91ZK0
Gene Alias activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; activating enhancer-binding protein 2-delta; AP-2 like; AP-2-delta; AP2-delta; hypothetical protein LOC566838; si:dkey-5e16.2; Tcfap2d; TFAP2BL1; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 beta-like 1; transcription factor AP-2 delta; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); transcription factor AP-2, delta; transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta; zgc:114196
Gene Symbols TFAP2D
Host Species Rabbit
Immunogen A synthesized peptide derived from human TFAP2D(Accession Q7Z6R9), corresponding to amino acid residues D303-S353.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226896, 301284, 83741
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.