Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFAP2E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541689 Supplier Invitrogen™ Supplier No. PA541689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FGGPAICAAL TAFQNYLLES LKGLDKMFLS SVGSGHGETK ASEKDAKHRK Sequence homology: Cow: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 100%.

TFAP2E is the sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. AP-2 epsilon may play a role in the development of the CNS and in cartilage differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFAP2E
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TFAP2E
Gene Accession No. Q6VUC0
Gene Alias activating enhancer-binding protein 2 epsilon; activating enhancer-binding protein 2-epsilon; adaptor-related protein complex 2, epsilon subunit; AI505200; AP2E; AP-2e; AP-2-epsilon; AP2-epsilon; hypothetical protein MGC65882; LOW QUALITY PROTEIN: transcription factor AP-2-epsilon; MGC49007; RGD1563297; Tcfap2e; Tfap2e; transcription factor AP-2 epsilon; transcription factor AP-2 epsilon (activating enhancer binding protein 2 epsilon); transcription factor AP-2, epsilon; transcription factor AP-2-epsilon; zgc:65882; zgc:65882 protein; zgc:77338
Gene Symbols TFAP2E
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TFAP2E (aa 393-442).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339488
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.