Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540720
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540720 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540720 Supplier Invitrogen™ Supplier No. PA540720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EEEQRRAVIV KPVRSCPEAP TSDTASDSEA SDSDAMDQSR EEPSGDGELP Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 93%.

Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFAP4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tfap4
Gene Accession No. Q01664
Gene Alias Activating enhancer-binding protein 4; AI642933; AP4; AP-4; bHLHc41; Class C basic helix-loop-helix protein 41; D930048N17Rik; LOW QUALITY PROTEIN: transcription factor AP-4; Tcfap4; TFAP4; transcription factor AP4; Transcription factor AP-4; transcription factor AP-4 (activating enhancer binding protein 4); transcription factor AP-4 (activating enhancer-binding protein 4)
Gene Symbols Tfap4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TFAP4 (aa 289-338).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7023
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.