Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFAP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 120171AP150UL

Description
Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNAbinding, and HLH and ZIP domains, which are used foroligomerization. Transcription factor AP4 activates both viral andcellular genes by binding to the symmetrical DNA sequence CAGCTG.Specifications
TFAP4 | |
Polyclonal | |
Unconjugated | |
TFAP4 | |
AP 4, bHLHc41, TFAP4, Transcription factor AP 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360482, 7023, 83383 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q01664 | |
Tfap4 | |
TFAP4 Fusion Protein Ag2633 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction