Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TFB-TBOA
GREENER_CHOICESDP

Catalog No. 25321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-321 1 mg
25-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-321 Supplier Tocris Bioscience™ Supplier No. 2532/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity EAAT1 and EAAT2 blocker

TFB-TBOA is a potent and selective glial glutamate transporter EAAT1 and EAAT2 inhibitor (IC50 values are 17, 22 and 300 nM for EAAT2, EAAT1 and EAAT3 respectively). Has no effect on EAAT4 and EAAT5, or a wide range of neuronal receptors and transporters. Attenuates glutamate-stimulated intracellular Na+ elevation in astrocytes in vitro (IC50 = 43 nM). Induces severe convulsions in vivo.

Specifications

Chemical Name or Material (3S)-3-[[3-[[4-(Trifluoromethyl)benzoyl]amino]phenyl]methoxy]-L-aspartic acid
CAS 480439-73-4
Assay Percent Range >98%
Quantity 1 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C19H17F3N2O6
Formula Weight Observed MW: 426.35

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.