Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFB1M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556142 Supplier Invitrogen™ Supplier No. PA556142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRSRLSVMAQ YLCNVRHIFT IPGQAFVPKP EVDVGVVHFT PLIQPKIEQP FKLVEKVVQN VFQFRRKYCH RGLRMLFPEA QRLESTGRLL ELADIDPTLR PRQLSISHFK SLCDVYRKMC DEDPQL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

The protein encoded by this gene is a dimethyltransferase that methylates the conserved stem loop of mitochondrial 12S rRNA. The encoded protein also is part of the basal mitochondrial transcription complex and is necessary for mitochondrial gene expression. The methylation and transcriptional activities of this protein are independent of one another. Variations in this gene may influence the severity of aminoglycoside-induced deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFB1M
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TFB1M
Gene Accession No. Q8WVM0
Gene Alias CGI75; CGI-75; Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; mtTFB; mtTFB1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1; TFB1M; transcription factor B1, mitochondrial
Gene Symbols TFB1M
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TFB1M. Recombinant protein control fragment (Product #RP-102224).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.