Learn More
Invitrogen™ TFB2M Polyclonal Antibody

Description
Immunogen sequence: GRFCILGSEAA TRKHLPARNH CGLSDSSPQL WPEPDFRNPP RKASKASLDF KRYVTDRRLA ETL Highest antigen sequence identity to the following orthologs - mouse 34%, rat 36%.
Specifications
Specifications
| Antigen | TFB2M |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Tfb2m |
| Gene Accession No. | Q9H5Q4 |
| Gene Alias | dimethyladenosine transferase 2, mitochondrial; FLJ22661; FLJ23182; HCV NS5A-transactivated protein 5; Hepatitis C virus NS5A-transactivated protein 5; Hkp1; h-mtTFB; h-mtTFB2; house-keeping protein 1; hTFB2M; mitochondrial 12S rRNA dimethylase 2; Mitochondrial transcription factor B2; mTFB2M; mtTFB2; NS5ATP5; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2; TFB2M; transcription factor B2, mitochondrial |
| Gene Symbols | Tfb2m |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.