Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TFC 007
GREENER_CHOICE

Catalog No. 510810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-081-0 10 mg
51-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-081-0 Supplier Tocris Bioscience™ Supplier No. 5108/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor

TFC 007 is a potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor (IC50 = 83 nM). Inhibits PGD2 production and late phase nasal blockage in an animal model of allergic rhinitis. Exhibits negligible effects on COX-1, COX-2, 5-LOX, LTC4 synthase, TXA2 synthase and microsomal prostaglandin E2 synthase.

Specifications

Quantity 10 mg
Product Type TFC 007
Purity >98%
Molecular Weight (g/mol) 487.55
Inhibitors TFC 007

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.