Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFCP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567091 Supplier Invitrogen™ Supplier No. PA567091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQQQQQKHEDG DSNGTFFVYH AIYL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 88%.

This gene encodes a transcription factor that binds the alpha-globin promoter and activates transcription of the alpha-globin gene. The encoded protein regulates erythroid gene expression, plays a role in the transcriptional switch of globin gene promoters, and it activates many other cellular and viral gene promoters. The gene product interacts with certain inflammatory response factors, and polymorphisms of this gene may be involved in the pathogenesis of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFCP2
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TFCP2
Gene Accession No. Q12800
Gene Alias alpha-globin transcription factor CP2; CP2; CP-2; D230015P20Rik; Late SV40 factor; LBP 1C; LBP1; LBP1C; LBP-1c; LBP-1d; LOW QUALITY PROTEIN: alpha-globin transcription factor CP2; LSF; LSF1D; SAA3 enhancer factor; SEF; Tcfcp2; TFCP2; TFCP2C; transcription factor CP2; transcription factor CP2 tv1; Transcription factor LSF; UBP-1
Gene Symbols TFCP2
Host Species Rabbit
Immunogen Recombinant Human TFCP2. Recombinant protein control fragment (Product #RP-105498).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.