Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFCP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576756
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a transcription factor that binds the alpha-globin promoter and activates transcription of the alpha-globin gene. The encoded protein regulates erythroid gene expression, plays a role in the transcriptional switch of globin gene promoters, and it activates many other cellular and viral gene promoters. The gene product interacts with certain inflammatory response factors, and polymorphisms of this gene may be involved in the pathogenesis of Alzheimer's disease.
Specifications
TFCP2 | |
Polyclonal | |
Unconjugated | |
TFCP2 | |
alpha-globin transcription factor CP2; CP2; CP-2; D230015P20Rik; Late SV40 factor; LBP 1C; LBP1; LBP1C; LBP-1c; LBP-1d; LOW QUALITY PROTEIN: alpha-globin transcription factor CP2; LSF; LSF1D; SAA3 enhancer factor; SEF; Tcfcp2; TFCP2; TFCP2C; transcription factor CP2; transcription factor CP2 tv1; Transcription factor LSF; UBP-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21422, 7024 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12800, Q9ERA0 | |
TFCP2 | |
Recombinant full length Human TFCP2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction