Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFCP2L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540451
Description
Peptide sequence: LRDVLALPIF KQEEPQLSPE NEARLPPLQY VLCAATSPAV KLHEETLTYL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Yeast: 75%.
TFCP2L1 is a candidate CP2 family member. It is expressed in a developmentally regulated fashion in vivo and acts as a direct repressor of transcription. CP2-related proteins comprise a family of DNA-binding transcription factors that are generally activators of transcription and expressed ubiquitously.
Specifications
TFCP2L1 | |
Polyclonal | |
Unconjugated | |
TFCP2L1 | |
1810030F05Rik; 4932442M07Rik; AA575098; Cp2l1; CP2-related transcriptional repressor 1; Crtr1; CRTR-1; D930018N21Rik; LBP9; LBP-9; Tcfcp2l1; Tcfcp2-related transcriptional repressor 1; Tfcp2l1; transcription factor CP2 like 1; transcription factor CP2-like 1; transcription factor CP2-like protein 1; Transcription factor LBP-9; transcription repressor CRTR-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29842 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZI6 | |
TFCP2L1 | |
Synthetic peptide directed towards the N-terminal of human TFCP2L1 (aa 20-69). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction