Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFCP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585976 Supplier Invitrogen™ Supplier No. PA585976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFCP2L1 Polyclonal Antibody for Western Blot

TFCP2L1 is a candidate CP2 family member. It is expressed in a developmentally regulated fashion in vivo and acts as a direct repressor of transcription. CP2-related proteins comprise a family of DNA-binding transcription factors that are generally activators of transcription and expressed ubiquitously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFCP2L1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% Glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene TFCP2L1
Gene Accession No. Q9NZI6
Gene Alias 1810030F05Rik; 4932442M07Rik; AA575098; Cp2l1; CP2-related transcriptional repressor 1; Crtr1; CRTR-1; D930018N21Rik; LBP9; LBP-9; Tcfcp2l1; Tcfcp2-related transcriptional repressor 1; Tfcp2l1; transcription factor CP2 like 1; transcription factor CP2-like 1; transcription factor CP2-like protein 1; Transcription factor LBP-9; transcription repressor CRTR-1
Gene Symbols TFCP2L1
Host Species Rabbit
Immunogen Synthetic peptide within Human TFCP2L1 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29842
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.