Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFCP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116028 Supplier Invitrogen™ Supplier No. PA5116028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TF2L1.

TFCP2L1 is a candidate CP2 family member. It is expressed in a developmentally regulated fashion in vivo and acts as a direct repressor of transcription. CP2-related proteins comprise a family of DNA-binding transcription factors that are generally activators of transcription and expressed ubiquitously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFCP2L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TFCP2L1
Gene Accession No. Q3UNW5, Q9NZI6
Gene Alias 1810030F05Rik; 4932442M07Rik; AA575098; Cp2l1; CP2-related transcriptional repressor 1; Crtr1; CRTR-1; D930018N21Rik; LBP9; LBP-9; Tcfcp2l1; Tcfcp2-related transcriptional repressor 1; Tfcp2l1; transcription factor CP2 like 1; transcription factor CP2-like 1; transcription factor CP2-like protein 1; Transcription factor LBP-9; transcription repressor CRTR-1
Gene Symbols TFCP2L1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TFCP2L1(Accession Q9NZI6), corresponding to amino acid residues M1-Q35.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29842, 304741, 81879
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.