Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFDP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145160 Supplier Invitrogen™ Supplier No. PA5145160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STVNPLGKQL LPKTFGQSSV NIDQQVVIG Antigen sequence identity: Mouse 90%; Rat 90%.

This gene encodes a member of the DP family of transcription factors. These factors heterodimerize with E2F proteins to enhance their DNA-binding activity and promote transcription from E2F target genes. This protein functions as a negative regulator and inhibits the DNA binding and transcriptional activities of E2F factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFDP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TFDP3
Gene Accession No. Q5H9I0
Gene Alias cancer/testis antigen 30; CT30; DP4; E2F-like protein; HCA661; Hepatocellular carcinoma-associated antigen 661; TFDP3; Transcription factor Dp family member 3; transcription factor Dp family, member 3
Gene Symbols TFDP3
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 32-60. Recombinant protein control fragment (Product #RP-110245).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.