Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFEB Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411680.025MG
Description
TFEB Polyclonal specifically detects TFEB in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TFEB | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
TFEB | |
Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TFEB. The immunogen is located within amino acids 390 - 440 of TFEB. Amino Acid Squence: FGGREDEGPPGYPEPL | |
Affinity Purified | |
RUO | |
7942 | |
Human | |
IgG |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg, Immunohistochemistry-Paraffin 2.5 μg/mL | |
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
Rabbit | |
53 kDa | |
0.025 mg | |
Primary | |
TFEB antibody is human specific. At least three isoforms of TFEB are known to exist. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction