Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFEB Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $512.00
Specifications
Antigen | TFEB |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
TFEB Polyclonal specifically detects TFEB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
TFEB | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
Polyclonal | |
Rabbit | |
Human | |
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
TFEB | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
7942 | |
Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human TFEB. The immunogen is located within amino acids 70 - 120 of TFEB. Amino Acid Squence: SQHQKVREYLSETYGNKF | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title