Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFEB Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212758F
Description
TFEB Polyclonal antibody specifically detects TFEB in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TFEB | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Transcription Factor EB using the numbering given in entry NP_009093.1 (GeneID 7942). | |
0.1 mL | |
Autophagy | |
7942 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction