Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFEB Recombinant Rabbit Monoclonal Antibody (BLR070G)

Catalog No. PIMA544332
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544332 20 μL
PIMA544331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA544332 Supplier Invitrogen™ Supplier No. MA544332
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

TFEB Recombinant Monoclonal Antibody for IP, Western Blot

The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFEB
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BLR070G
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene TFEB
Gene Accession No. P19484, Q9R210
Gene Alias Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB
Gene Symbols TFEB
Host Species Rabbit
Immunogen Peptide representing a region between residue 426 and the C-terminus (residue 476) of human Transcription factor EB.
Purification Method Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21425, 7942
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.