Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TFF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580110
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Stomach Tissue. IHC: Rat Intestine Tissue.
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.
Specifications
TFF1 | |
Polyclonal | |
Unconjugated | |
TFF1 | |
Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117270, 21784 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q08423, Q63467 | |
TFF1 | |
A synthetic peptide corresponding to a sequence in the middle region of mouse Estrogen Inducible Protein pS2 (59-72aa CCFDDSVRGFPWCF). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction