Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals TFF1/pS2 Antibody (GE2 (same as R47/94)) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2346230.1MG
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOFSpecifications
TFF1/pS2 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.5-1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 0.5-1 μg/mL, CyTOF-ready | |
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
A synthetic peptide (around aa 57-84) of human TFF1/pS2 polypeptide (exact sequence is proprietary) (Uniprot: P04155) | |
0.1 mg | |
Breast Cancer, Cancer | |
7031 | |
Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
GE2 (same as R47/94) | |
Unconjugated | |
10 mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction