Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFF1/pS2 Antibody (rTFF1/1091) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308248
Description
TFF1/pS2 Monoclonal specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
TFF1/pS2 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
Recombinant full-length human TFF1/pS2 protein | |
100 μg | |
Breast Cancer, Cancer | |
7031 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rTFF1/1091 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction