Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFF1/pS2 Antibody (TFF1/2969R), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308252DL594
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human, Cynomolgus Monkey samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TFF1/pS2 | |
Monoclonal | |
DyLight 594 | |
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human TFF1/pS2 (Uniprot: P04155) | |
100 μg | |
Breast Cancer, Cancer | |
7031 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
TFF1/2969R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction