Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFF3 Antibody (SR2317), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321814100UL
Description
TFF3 Monoclonal antibody specifically detects TFF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
TFF3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR2317 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:30 | |
HITF, human intestinal trefoil factor10Intestinal trefoil factor, trefoil factor 3 (intestinal) | |
A synthesized peptide derived from human TFF3 (Uniprot #: Q07654) | |
100 μg | |
Biologically Active Proteins, Cancer | |
7033 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction